Please login to your online account to display your discounted pricing

CD40 Mouse anti-Human, PE, Clone: 5C3, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $317.55

Specifications

Antigen CD40
Clone 5C3
Host Species Mouse
Gene Alias TNFRSF5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010353

 
affymetrix
12040941
25 tests Each for $129.05

5010354

 
affymetrix
12040942
100 tests Each for $317.55
Description & Specifications

Specifications

Antigen CD40
Clone 5C3
Host Species Mouse
Gene Alias TNFRSF5
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.015μg)/test

The 5C3 monoclonal antibody reacts with human CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development and maturation by inducing Ig isotype-switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation. 5C3 is reported to be used for activation of B cells in in vitro functional assays.