missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD40, eFluor 450, clone: HM40-3, eBioscience™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Life Technologies LS48040280

 View more versions of this product

Catalog No. 501129283



Description

Description

The CD40 antigen is a single chain glycoprotein. It is known to be a member of the tumor necrosis factor / nerve growth factor superfamily and shows a significant homology to the Hodgkin's disease associated antigen, CD30. CD40 is present on all B cells except plasma cells. It is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. The precise function of the CD40 antigen is unknown but it appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.

Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered. The CD40 antigen is a single chain glycoprotein that is known to be a member of the tumor necrosis factor/nerve growth factor superfamily and shows a significant homology to the Hodgkin′s disease associated antigen, CD30. CD40 is present on all B cells except plasma cells and is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. CD40 has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. AT-hook transcription factor AKNA is reported to coordinately regulate the expression of CD40, which may be important for homotypic cell interactions. Adaptor protein TNFR2 interacts with CD40 and serves as a mediator of the signal transduction. The interaction of CD40 and its ligand is found to be necessary for amyloid-beta-induced microglial activation, and thus is thought to be an early event in Alzheimer disease pathogenesis. Two alternatively spliced transcript variants of CD40 encoding distinct isoforms have been reported. Diseases associated with CD40 dysfunction include Type 3 Hyper-Igm immunodeficiency and CD40 ligand deficiency.
Specifications

Specifications

CD40
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P27512
Bp50, CDW40, MGC9013, TNFRSF5, p50
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse, Rat
Flow Cytometry
HM40-3
eFluor® 450
Cd40
Liquid
Cd40
IgM, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
171369, 21939
Documents

For Research Use Only.