missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD40 Armenian Hamster anti-Mouse, Rat, APC, Clone: HM40-3, eBioscience™

Armenian Hamster Monoclonal Antibody

$117.48 - $315.48

Specifications

Antigen CD40
Clone HM40-3
Host Species Armenian Hamster
Gene Alias TNFRSF5
Species Reactivity Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245644 Life Technologies
LS17040280
25μg Each for $117.48
50245645 Life Technologies
LS17040282
100μg Each for $315.48
Specifications

Description

The HM40-3 monoclonal antibody reacts with mouse and rat CD40, a 45-50kDa type I transmembrane glycoprotein. CD40 is a member of the TNFR family and is expressed by mouse B lymphocytes, follicular dendritic cells, thymic epithelium, and a subset of peripheral T cells. CD40 regulates B cell development/maturation by inducing Ig isotype switching and in combination with other signals such as IL-4, protects B cells from surface Ig-induced apoptosis and promotes proliferation. Interaction of CD40 with CD154 (gp39), its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.
Description & Specifications

Specifications

CD40
Armenian Hamster
Mouse, Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HM40-3
TNFRSF5
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL