Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, PE-Cy5, Clone: GK1.5, eBioscience™

Rat Monoclonal Antibody

$191.40 - $590.15

Specifications

Antigen CD4
Clone GK1.5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013995

 
life technologies
LS15004181
50μg Each for $191.40

5013996

 
life technologies
LS15004182
100μg Each for $292.90

5013997

 
life technologies
LS15004183
200μg Each for $590.15
Description & Specifications

Specifications

Antigen CD4
Clone GK1.5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The GK1.5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of GK1.5 is blocked by RM4-5.