Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, PE, Clone: RM4-5, eBioscience™

Rat Monoclonal Antibody

$100.05 - $462.55

Specifications

Antigen CD4
Clone RM4-5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010157

 
life technologies
LS12004281
50μg Each for $100.05

5010159

 
life technologies
LS12004283
200μg Each for $230.55

5010158

 
life technologies
LS12004282
100μg Each for $143.55

5010160

 
life technologies
LS12004285
500μg Each for $462.55
Description & Specifications

Specifications

Antigen CD4
Clone RM4-5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The RM4-5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of RM4-5 is blocked by GK1.5.