missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, FITC, Clone: RM4-5, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS11004282

 View more versions of this product

Catalog No. 501129681



Description

Description

Description: The RM4-5 monoclonal antibody reacts with the mouse CD4 molecule, a 55 kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of RM4-5 is blocked by GK1.5. Applications Reported: The RM4-5 antibody has been reported for use in flow cytometric analysis. Applications Tested: The RM4-5 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
Specifications

Specifications

CD4
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06332
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
RM4-5
FITC
Cd4
Liquid
Cd4
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12504
Documents

For Research Use Only.