Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, eFluor 660, Clone: 4SM95, eBioscience™

Rat Monoclonal Antibody

$175.45 - $427.75

Specifications

Antigen CD4
Clone 4SM95
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124533

 
life technologies
LS50976680
25μg Each for $175.45

501124534

 
life technologies
LS50976682
100μg Each for $427.75
Description & Specifications

Specifications

Antigen CD4
Clone 4SM95
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This 4SM95 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T cells, and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail.

The 4SM95 monoclonal antibody is not recommended for immunohistochemistry of frozen mouse tissue. For detection of mouse CD4 using immunohistochemistry of frozen tissue please refer to alternate products Anti-Mouse CD4 Purified (cat. 14-0041 and 14-0042).