missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4, eFluor 660, clone: 4SM95, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS50976680

 View more versions of this product

Catalog No. 501124533



Description

Description

The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].

Description: This 4SM95 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T cells, and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail.The 4SM95 monoclonal antibody is not recommended for immunohistochemistry of frozen mouse tissue. For detection of mouse CD4 using immunohistochemistry of frozen tissue please refer to alternate products Anti-Mouse CD4 Purified (cat. 14-0041 and 14-0042).Applications Reported: This 4SM95 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy.Applications Tested: This 4SM95 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded mouse tissue using low pH antigen retrieval and can be used at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD4
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P06332
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
4SM95
eFluor 660
Cd4
Liquid
Cd4
IgG1, kappa, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12504
Documents

For Research Use Only.