missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, eFluor 450, Clone: GK1.5, eBioscience™

Rat Monoclonal Antibody

$145.00 - $324.80

Specifications

Antigen CD4
Clone GK1.5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016234 Life Technologies
LS48004180
25μg Each for $145.00
5016235 Life Technologies
LS48004182
100μg Each for $324.80
Specifications

Description

The GK1.5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of GK1.5 is blocked by RM4-5.
Description & Specifications

Specifications

CD4
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
GK1.5
L3T4, Ly-4
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL