missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, Clone: RM4-5, eBioscience™

Rat Monoclonal Antibody

$64.68 - $447.48

Specifications

Antigen CD4
Clone RM4-5
Host Species Rat
Gene Alias L3T4, Ly-4
Applications Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012363 Life Technologies
LS14004281
50μg Each for $64.68
5012364 Life Technologies
LS14004282
100μg Each for $117.48
5012365 Life Technologies
LS14004285
500μg Each for $249.48
5012366 Life Technologies
LS14004286
1mg Each for $447.48
Specifications

Description

The RM4-5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of RM4-5 is blocked by GK1.5.
Description & Specifications

Specifications

CD4
Rat
Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
RM4-5
L3T4, Ly-4
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer