Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, Clone: GK1.5, Functional Grade, eBioscience™

Rat Monoclonal Antibody

$114.55 - $3,623.55

Specifications

Antigen CD4
Clone GK1.5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122936

 
affymetrix
16004195
10mg Each for $3,623.55

5014198

 
affymetrix
16004182
100μg Each for $172.55

5014197

 
affymetrix
16004181
50μg Each for $114.55

5014200

 
affymetrix
16004186
1mg Each for $607.55

5014199

 
affymetrix
16004185
500μg Each for $332.05
Description & Specifications

Specifications

Antigen CD4
Clone GK1.5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide

The GK1.5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of GK1.5 is blocked by RM4-5.