Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, Biotin, Clone: RM4-5, eBioscience™

Rat Monoclonal Antibody

$85.55 - $564.05

Specifications

Antigen CD4
Clone RM4-5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011388

 
life technologies
LS13004281
50μg Each for $85.55

5011389

 
life technologies
LS13004282
100μg Each for $143.55

5011390

 
life technologies
LS13004285
500μg Each for $332.05

5011391

 
life technologies
LS13004286
1mg Each for $564.05
Description & Specifications

Specifications

Antigen CD4
Clone RM4-5
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The RM4-5 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with protein tyrosine kinase p56lck through its cytoplasmic tail. Binding of RM4-5 is blocked by GK1.5.