missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Rat anti-Mouse, Biotin, Clone: RM4-4, eBioscience™

Rat Monoclonal Antibody

$85.55 - $332.05

Specifications

Antigen CD4
Clone RM4-4
Host Species Rat
Gene Alias L3T4, Ly-4
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011392 Life Technologies
LS13004381
50μg Each for $85.55
5011393 Life Technologies
LS13004382
100μg Each for $143.55
5011394 Life Technologies
LS13004385
500μg Each for $332.05
Specifications

Description

The RM4-4 monoclonal antibody reacts with the mouse CD4 molecule, a 55kDa cell surface receptor expressed by the majority of thymocytes, a subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with the protein tyrosine kinase lck through its cytoplasmic tail. Binding of RM4-4 does not block binding of the CD4 monoclonal antibodies RM4-5 or GK1.5.
Description & Specifications

Specifications

CD4
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RM4-4
L3T4, Ly-4
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL