Please login to your online account to display your discounted pricing

CD4 Rat anti-Canine, PE-Cy7, Clone: YKIX302.9, eBioscience™

Rat Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen CD4
Clone YKIX302.9
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123377

 
affymetrix
25504041
25 tests Each for $158.05

501129730

 
affymetrix
25504042
100 tests Each for $390.05
Description & Specifications

Specifications

Antigen CD4
Clone YKIX302.9
Host Species Rat
Species Reactivity Canine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The YKIX302.9 monoclonal antibody reacts with canine CD4, a cell surface receptor expressed on a majority of thymocytes, a subpopulation of mature T cells and dendritic cells. In dogs, CD4 is also highly expressed on neutrophils. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role in T cell development and homeostasis.

This YKIX302.9 monoclonal antibody has been reported to block MHC II-restricted T cell responses in vivo and in vitro.