Please login to your online account to display your discounted pricing

CD4 Mouse anti-Rat, FITC, Clone: OX35, eBioscience™

Mouse Monoclonal Antibody

$100.05 - $375.55

Specifications

Antigen CD4
Clone OX35
Host Species Murine
Gene Alias W3/25
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509373

 
life technologies
LS11004081
50μg Each for $100.05

509374

 
life technologies
LS11004082
100μg Each for $162.40

509375

 
life technologies
LS11004085
500μg Each for $375.55
Description & Specifications

Specifications

Antigen CD4
Clone OX35
Host Species Murine
Gene Alias W3/25
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The OX35 monoclonal antibody reacts with rat CD4. CD4 is a co-receptor which participates in signaling through the T cell receptor by making contacts with MHC class II expressed on antigen-presenting cells (APC). CD4 is expressed in the thymus by CD4+CD8+ “double-positive” and CD4+ “single-positive” thymocytes, and by CD4+ “helper” T cells in the periphery. CD4 is a cell-surface membrane glycoprotein which contains four extracellular immunoglobulin-like domains.