missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Mouse anti-Rat, APC, Clone: OX35, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $341.88

Specifications

Antigen CD4
Clone OX35
Host Species Murine
Gene Alias W3/25
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014848 Life Technologies
LS17004080
25μg Each for $117.48
5014849 Life Technologies
LS17004082
100μg Each for $341.88
Specifications

Description

The OX35 monoclonal antibody reacts with rat CD4. CD4 is a co-receptor which participates in signaling through the T cell receptor by making contacts with MHC class II expressed on antigen-presenting cells (APC). CD4 is expressed in the thymus by CD4+CD8+ “double-positive” and CD4+ “single-positive” thymocytes, and by CD4+ “helper” T cells in the periphery. CD4 is a cell-surface membrane glycoprotein which contains four extracellular immunoglobulin-like domains.
Description & Specifications

Specifications

CD4
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
OX35
W3/25
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer