missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, FITC, Clone: SK3 (SK-3), eBioscience™

Mouse Monoclonal Antibody

$172.55 - $346.55

Specifications

Antigen CD4
Clone SK3 (SK-3)
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509388 Life Technologies
LS11004741
25 Tests Each for $172.55
509389 Life Technologies
LS11004742
100 Tests Each for $346.55
Specifications

Description

The SK3 monoclonal antibody reacts with human CD4, a 59kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and, in low levels, monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). SK3 blocks HIV binding and mixed lymphocyte reaction. The SK3 and RPA-T4 monoclonal antibodies do not cross-block binding, suggesting recognition of distinct epitopes.
Description & Specifications

Specifications

CD4
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SK3 (SK-3)
Leu-3, T4
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test