Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, eVolve 655, Clone: SK3 (SK-3), eBioscience™

Mouse Monoclonal Antibody

$261.00 - $437.90

Specifications

Antigen CD4
Clone SK3 (SK-3)
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125038

 
life technologies
LS86004741
25 Tests Each for $261.00

501125039

 
life technologies
LS86004742
100 Tests Each for $437.90
Description & Specifications

Specifications

Antigen CD4
Clone SK3 (SK-3)
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eVolve 655
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL

The SK3 monoclonal antibody reacts with human CD4, a 59kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and at low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). SK3 blocks HIV binding and mixed lymphocyte reaction. The SK3 and RPA-T4 monoclonal antibodies do not cross-block binding, suggesting recognition of distinct epitopes.