Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, eFluor 570, Clone: N1UG0, eBioscience™

Mouse Monoclonal Antibody

$146.45 - $368.30

Specifications

Antigen CD4
Clone N1UG0
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123597

 
life technologies
LS41244480
25μg Each for $146.45

501123598

 
life technologies
LS41244482
100μg Each for $368.30
Description & Specifications

Specifications

Antigen CD4
Clone N1UG0
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Regulatory Status RUO
Conjugate eFluor 570
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The N1UG0 monoclonal antibody reacts with human CD4, a 59kDa cell surface glycoprotein expressed by the majority of thymocytes, a subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The N1UG0 antibody is recommended for use in staining human formalin-fixed paraffin embedded tissue sections.