Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, eFluor 450, Clone: OKT4 (OKT-4), eBioscience™

Mouse Monoclonal Antibody

$172.55 - $368.30

Specifications

Antigen CD4
Clone OKT4 (OKT-4)
Host Species Mouse
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016240

 
life technologies
LS48004841
25 tests Each for $172.55

5016241

 
life technologies
LS48004842
100 tests Each for $368.30
Description & Specifications

Specifications

Antigen CD4
Clone OKT4 (OKT-4)
Host Species Mouse
Gene Alias Leu-3, T4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The OKT4 monoclonal antibody reacts with human CD4, a 59kDa cell surface glycoprotein expressed by the majority of thymocytes, a subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The OKT4 antibody recognizes a different epitope than the RPA-T4 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes.