Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, Clone: OKT4 (OKT-4), Functional Grade, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $477.05

Specifications

Antigen CD4
Clone OKT4 (OKT-4)
Host Species Mouse
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014206

 
life technologies
LS16004881
50μg Each for $158.05

5014207

 
life technologies
LS16004885
500μg Each for $477.05
Description & Specifications

Specifications

Antigen CD4
Clone OKT4 (OKT-4)
Host Species Mouse
Gene Alias Leu-3, T4
Species Reactivity Human
Applications Flow Cytometry
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide

The OKT4 monoclonal antibody reacts with human CD4, a 59kDa cell surface glycoprotein expressed by the majority of thymocytes, a subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The OKT4 antibody recognizes a different epitope than the RPA-T4 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes.