missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, Clone: N1UG0, eBioscience™

Mouse Monoclonal Antibody

$92.40 - $227.04

Specifications

Antigen CD4
Clone N1UG0
Host Species Murine
Gene Alias Leu-3, T4
Applications Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122665 Life Technologies
LS14244482
100μg Each for $227.04
501122664 Life Technologies
LS14244480
25μg Each for $92.40
Specifications

Description

The N1UG0 monoclonal antibody reacts with human CD4, a 59kDa cell surface glycoprotein expressed by the majority of thymocytes, a subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The N1UG0 antibody is recommended for use in staining human formalin-fixed paraffin embedded tissue sections.
Description & Specifications

Specifications

CD4
Murine
Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
N1UG0
Leu-3, T4
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer