Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, Biotin, Clone: RPA-T4, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD4
Clone RPA-T4
Host Species Mouse
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011398

 
life technologies
LS13004980
25μg Each for $114.55

501129379

 
life technologies
LS13004982
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD4
Clone RPA-T4
Host Species Mouse
Gene Alias Leu-3, T4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The RPA-T4 monoclonal antibody reacts with human CD4, a 59kDa cell surface receptor expressed by a majority of thymocytes, subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). RPA-T4 blocks HIV binding and mixed lymphocyte reaction. The RPA-T4 antibody recognizes a different epitope than the OKT4 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes.