missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, APC-eFluor 780, Clone: OKT4 (OKT-4), eBioscience™

Mouse Monoclonal Antibody

$158.05 - $427.75

Specifications

Antigen CD4
Clone OKT4 (OKT-4)
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124002 Life Technologies
LS47004841
25 Tests Each for $158.05
501128685 Life Technologies
LS47004842
100 Tests Each for $427.75
Specifications

Description

The OKT4 monoclonal antibody reacts with human CD4, a 59kDa cell surface glycoprotein expressed by the majority of thymocytes, a subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The OKT4 antibody recognizes a different epitope than the RPA-T4 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes.
Description & Specifications

Specifications

CD4
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OKT4 (OKT-4)
Leu-3, T4
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.125μg)/test