Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, APC, Clone: SK3, eBioscience™

Mouse Monoclonal Antibody

$230.55 - $448.05

Specifications

Antigen CD4
Clone SK3 (SK-3)
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014856

 
life technologies
LS17004741
25 Tests Each for $230.55

5014857

 
life technologies
LS17004742
100 Tests Each for $448.05
Description & Specifications

Specifications

Antigen CD4
Clone SK3 (SK-3)
Host Species Murine
Gene Alias Leu-3, T4
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The SK3 monoclonal antibody reacts with human CD4, a 59kDa cell surface receptor expressed by a majority of thymocytes, a subpopulation of mature T helper cells, and, in low levels, monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). SK3 blocks HIV binding and mixed lymphocyte reaction. The SK3 and RPA-T4 monoclonal antibodies do not cross-block binding, suggesting recognition of distinct epitopes.