missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD4 Mouse anti-Human, APC, Clone: OKT4, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $341.88

Specifications

Antigen CD4
Clone OKT4 (OKT-4)
Host Species Murine
Gene Alias Leu-3, T4
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014858 Life Technologies
LS17004841
25 Tests Each for $157.08
5014859 Life Technologies
LS17004842
100 Tests Each for $341.88
Specifications

Description

The OKT4 monoclonal antibody reacts with human CD4, a 59kDa cell surface glycoprotein expressed by the majority of thymocytes, a subpopulation of mature T cells (T-helper cells) and in low levels on monocytes. CD4 is a receptor for the human immunodeficiency virus (HIV). The OKT4 antibody recognizes a different epitope than the RPA-T4 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes.
Description & Specifications

Specifications

CD4
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test
OKT4 (OKT-4)
Leu-3, T4
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer