Please login to your online account to display your discounted pricing

CD4, CD25 anti-Human, FITC, PE, Clone: RPA-T4 (CD4) and BC96 (CD25), eZfluor™, Antibody Cocktail, eBioscience™

Monoclonal Antibody Cocktail

$230.55 - $564.05

Specifications

Antigen CD4, CD25
Clone RPA-T4 (CD4) and BC96 (CD25)
Gene Alias CD4 (RPA-T4); CD25 (BC96)
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015421

 
life technologies
LS22842571
25 tests Each for $230.55

5015422

 
life technologies
LS22842573
100 tests Each for $564.05
Description & Specifications

Specifications

Antigen CD4, CD25
Clone RPA-T4 (CD4) and BC96 (CD25)
Gene Alias CD4 (RPA-T4); CD25 (BC96)
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC, PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 20μL/test

This product consists of two fluorochrome-conjugated monoclonal antibodies to identify CD4+CD25+ human peripheral blood cells. The RPA-T4 monoclonal antibody (mouse IgG1, κ) reacts with human CD4, a 59 kDa cell surface receptor that is expressed by a majority of thymocytes, a subpopulation of mature T cells (T helper cells) and at low levels on monocytes. The BC96 monoclonal antibody (mouse IgG1, κ) reacts with human CD25, the 55 kDa interleukin-2 receptor α chain (IL-2Rα). This cocktail of RPA-T4 and BC96 can be used for the flow cytometric detection of human regulatory T cells, which also express Foxp3.