missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD3e Golden Syrian Hamster anti-Mouse, PerCP-eFluor 710, Clone: eBio500A2 (500A2), eBioscience™

Golden Syrian Hamster Monoclonal Antibody

$172.55 - $477.05

Specifications

Antigen CD3e
Clone eBio500A2 (500A2)
Host Species Golden Syrian Hamster
Gene Alias CD3 epsilon
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015893 Life Technologies
LS46003380
25μg Each for $172.55
5015894 Life Technologies
LS46003382
100μg Each for $477.05
Specifications

Description

The eBio500A2 monoclonal antibody reacts with the 25 kD epsilon subunit of the mouse CD3 complex. CD3 subunits γ, δ and ε are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, as well as NKT cells. Binding of eBio500A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation. The 500A2 antibody is able to partially cross-block the 17A2 and 145-2C11 antibodies, indicating that all three of these anti-CD3 antibodies recognize distinct but overlapping epitopes.
Description & Specifications

Specifications

CD3e
Golden Syrian Hamster
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio500A2 (500A2)
CD3 epsilon
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL