Please login to your online account to display your discounted pricing

CD3e Golden Syrian Hamster anti-Mouse, Alexa Fluor 700, Clone: 500A2, eBioscience™

Golden Syrian Hamster Monoclonal Antibody

$172.55 - $172.55

Specifications

Antigen CD3e
Clone eBio500A2 (500A2)
Host Species Golden Syrian Hamster
Gene Alias CD3 epsilon
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016843

 
affymetrix
56003380
25μg Each for $172.55
Description & Specifications

Specifications

Antigen CD3e
Clone eBio500A2 (500A2)
Host Species Golden Syrian Hamster
Gene Alias CD3 epsilon
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 700
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBio500A2 monoclonal antibody reacts with the 25 kD epsilon subunit of the mouse CD3 complex. CD3 subunits γ, δ and ε are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, as well as NKT cells. Binding of eBio500A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation. The 500A2 antibody is able to partially cross-block the 17A2 and 145-2C11 antibodies, indicating that all three of these anti-CD3 antibodies recognize distinct but overlapping epitopes.