Please login to your online account to display your discounted pricing

CD3e Armenian Hamster anti-Mouse, PE-Cy5, Clone: 145-2C11, eBioscience™

Armenian Hamster Monoclonal Antibody

$56.55 - $520.55

Specifications

Antigen CD3e
Clone 145-2C11
Host Species Armenian Hamster
Gene Alias CD3 epsilon
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122901

 
life technologies
LS15003181
50μg Each for $116.00

5013989

 
life technologies
LS15003163
10μg Each for $56.55

5013992

 
life technologies
LS15003183
200μg Each for $520.55

501129404

 
life technologies
LS15003182
100μg Each for $274.05
Description & Specifications

Specifications

Antigen CD3e
Clone 145-2C11
Host Species Armenian Hamster
Gene Alias CD3 epsilon
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 145-2C11 monoclonal antibody reacts with mouse CD3e, a 20kDa subunit of the TCR complex. Along with the other CD3 subunits, gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 145-2C11 to TCR initiates the intracellular biochemical pathway resulting in cellular activation, proliferation, and apoptosis depending on specific conditions utilized. 145-2C11 is commonly used as a phenotypic marker for mouse T cells.