missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD39 Rat anti-Mouse, PE-Cy7, Clone: 24DMS1, eBioscience™

Rat Monoclonal Antibody

$265.35 - $516.20

Specifications

Antigen CD39
Clone 24DMS1
Host Species Rat
Gene Alias Ectonucleoside Triphosphate Diphosphohydrolase 1, Entpd1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015505 Life Technologies
LS25039180
25μg Each for $265.35
501128745 Life Technologies
LS25039182
100μg Each for $516.20
Specifications

Description

The 24DMS1 monoclonal antibody reacts with mouse CD39, also known as NTPDaseI. E-NTPDases are enzymes that convert nucleoside tri- and diphosphates (NTDPs) into nucleoside monophosphate (NMP), thereby removing toxic extracellular ATP and ADP. CD39 is the dominant member of this family in the immune system and is involved in suppression of inflammation and control of platelet activation. CD39 can impact expression of CD73, another E-NTPase. Together, these molecules influence inflammation responses. CD39 is expressed on B cells, Langerhans cells and most monocytes. In addition, CD39 is found on a subset of CD4+ T cells that are mostly CD25+FoxP3+ T reg cells. T reg cells from CD39-null mice showed impaired suppressive properties in vitro and in vivo.
Description & Specifications

Specifications

CD39
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
24DMS1
Ectonucleoside Triphosphate Diphosphohydrolase 1, Entpd1
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL