missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD38 Rat anti-Mouse, APC, Clone: 90, eBioscience™

Rat Monoclonal Antibody

$227.04 - $402.60

Specifications

Antigen CD38
Clone 90
Host Species Rat
Gene Alias ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123048 Life Technologies
LS17038182
100μg Each for $402.60
501128883 Life Technologies
LS17038181
50μg Each for $227.04
Specifications

Description

The 90 monoclonal antibody reacts with the mouse CD38 molecule, an ∽42kDa type II transmembrane protein. CD38 is expressed at increasingly higher levels on B cells at each stage of B-cell differentiation, and is then down-regulated on germinal center B cells and mature plasma cells. Its expression is reported on a subpopulation of thymocytes, mature T cells, and NK cells. Crosslinking of CD38 on the surface of mature, resting B cells induces B-cell proliferation, which is enhanced by co-signals such as IL-4 and LPS. CD38, a counter-receptor for CD31, is an ectoenzyme with cyclase and hydrolase enzymatic activity and is speculated to play a role in lymphocyte activation and differentiation.
Description & Specifications

Specifications

CD38
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
90
ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL