Please login to your online account to display your discounted pricing

CD38 Mouse anti-Human, PE-eFluor 610, Clone: HIT2, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen CD38
Clone HIT2
Host Species Murine
Gene Alias ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124755

 
life technologies
LS61038941
25 Tests Each for $172.55

501124756

 
life technologies
LS61038942
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen CD38
Clone HIT2
Host Species Murine
Gene Alias ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The HIT2 monoclonal antibody reacts with the human CD38 molecule, an approximately 45kDa type II transmembrane protein. CD38 is expressed by thymocytes, peripheral B cells including plasma cells, activated T cells, and monocytes. CD38 is a counter-receptor for CD31 and has both cyclase and hydrolase enzymatic activity.