missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD38 Mouse anti-Human, PE-Cy7, Clone: HIT2, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $442.25

Specifications

Antigen CD38
Clone HIT2
Host Species Murine
Gene Alias ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015503 Life Technologies
LS25038941
25 Tests Each for $216.05
5015504 Life Technologies
LS25038942
100 Tests Each for $442.25
Specifications

Description

The HIT2 monoclonal antibody reacts with the human CD38 molecule, an approximately 45kDa type II transmembrane protein. CD38 is expressed by thymocytes, peripheral B cells including plasma cells, activated T cells, and monocytes. CD38 is a counter-receptor for CD31 and has both cyclase and hydrolase enzymatic activity.
Description & Specifications

Specifications

CD38
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HIT2
ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (1μg)/test