Please login to your online account to display your discounted pricing

CD38 Mouse anti-Human, eFluor 450, Clone: HIT2, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $442.25

Specifications

Antigen CD38
Clone HIT2
Host Species Mouse
Gene Alias ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124109

 
life technologies
LS48038941
25 tests Each for $216.05

501124110

 
life technologies
LS48038942
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen CD38
Clone HIT2
Host Species Mouse
Gene Alias ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The HIT2 monoclonal antibody reacts with the human CD38 molecule, an approximately 45kDa type II transmembrane protein. CD38 is expressed by thymocytes, peripheral B cells including plasma cells, activated T cells, and monocytes. CD38 is a counter-receptor for CD31 and has both cyclase and hydrolase enzymatic activity.