Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD37 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD37 Ab-1, Mouse Monoclonal Antibody.

$468.06 - $1,028.16

Specifications

Antigen CD37 Ab-1
Clone IPO-24
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS541P

 
thermo scientific™ lab vision™
MS541P
200μg/mL 1mL Each for $924.18

MS541P0

 
thermo scientific™ lab vision™
MS541P0
200μg/mL 100µL Each for $468.06

MS541P1

 
thermo scientific™ lab vision™
MS541P1
200μg/mL 500µL Each for $835.24

MS541P1ABX

 
thermo scientific™ lab vision™
MS541P1ABX
1mg/mL 100µL Each for $956.82

MS541PABX

 
thermo scientific™ lab vision™
MS541PABX
1mg/mL 200µL Each for $1,028.16
Description & Specifications

Specifications

Antigen CD37 Ab-1
Clone IPO-24
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline B-Cell
Type Primary
Immunogen Spleen cells of a patient with hairy cell leukemia
Isotype IgG2b kappa

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD37 Ab-1, Mouse Monoclonal Antibody.

CD37, a tetraspan transmembrane protein, is strongly expressed on normal and neoplastic mature (sIg+) B lymphocytes. In B-cell ontogeny, CD37 appears after the pre-B-cell stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD37 is absent from platelets and erythrocytes. It associates with other tetraspan transmembrane proteins and MHC class II molecules to form a large complexes at the surface of B cells and play a role in signal transduction. CD37 is a valuable and stable marker for peripheral mature B-cells and corresponding malignancies like B-cell chronic lymphocytic leukemia (B-CLL), hairy cell leukemia (HCL), and all types of B-cell non-Hodgkin lymphoma (B-NHL).

Host Species: Mouse

Clone: IPO-24

Isotype: IgG2b/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Spleen cells of a patient with hairy cell leukemia

Molecular Weight: 33-55 kDa

Positive Control: Raji cells, Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen only)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO