Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD37 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD37 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS541P1ABX

 View more versions of this product

Catalog No. MS541P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen Spleen cells of a patient with hairy cell leukemia
Regulatory Status RUO
Clone IPO-24
Isotype IgG2b kappa
Cross Reactivity Human
Antigen CD37 Ab-1
Format Concentrated
Host Species Mouse
Quantity 100µL
Classification Monoclonal
Research Discipline B-Cell
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD37 Ab-1, Mouse Monoclonal Antibody.

CD37, a tetraspan transmembrane protein, is strongly expressed on normal and neoplastic mature (sIg+) B lymphocytes. In B-cell ontogeny, CD37 appears after the pre-B-cell stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD37 is absent from platelets and erythrocytes. It associates with other tetraspan transmembrane proteins and MHC class II molecules to form a large complexes at the surface of B cells and play a role in signal transduction. CD37 is a valuable and stable marker for peripheral mature B-cells and corresponding malignancies like B-cell chronic lymphocytic leukemia (B-CLL), hairy cell leukemia (HCL), and all types of B-cell non-Hodgkin lymphoma (B-NHL).

Host Species: Mouse

Clone: IPO-24

Isotype: IgG2b/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Spleen cells of a patient with hairy cell leukemia

Molecular Weight: 33-55 kDa

Positive Control: Raji cells, Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen only)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO