missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD368 (Clec6) Mouse anti-Human, eFluor 660, Clone: 9B9, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $421.08

Specifications

Antigen CD368 (Clec6)
Clone 9B9
Host Species Murine
Gene Alias CLEC4D, CLECSF8
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124495 Life Technologies
LS50911341
25 Tests Each for $170.28
501124496 Life Technologies
LS50911342
100 Tests Each for $421.08
Specifications

Description

This 9B9 monoclonal antibody reacts with human CD368, also known as Clec6 or ClecsF8 or Clec4D. In peripheral blood, this C-type lectin is expressed highly on granulocytes and CD14+CD16- monocytes. Some dendritic cell populations also exhibit low levels of CD368 expression. Studies have demonstrated that this receptor undergoes endocytosis due to its C-type lectin-like domain. CD368 functions as a phagocytic receptor and is involved in proinflammatory cytokine production in cooperation with Toll-like receptors. Finally, CD368 also participates in cellular activation via signaling through Syk kinase.
Description & Specifications

Specifications

CD368 (Clec6)
Murine
Flow Cytometry
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test
9B9
CLEC4D, CLECSF8
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer