missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD366 (TIM3) Rat anti-Mouse, Biotin, Clone: 8B.2C12, eBioscience™

Rat Monoclonal Antibody

$172.55 - $680.05

Specifications

Antigen CD366 (TIM3)
Clone 8B.2C12
Host Species Rat
Gene Alias TIM-3, HAVCR2, T cell immunoglobulin domain, mucin-like domain
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011959 Life Technologies
LS13587181
50μg Each for $172.55
5011960 Life Technologies
LS13587182
100μg Each for $317.55
5011961 Life Technologies
LS13587185
500μg Each for $680.05
Specifications

Description

The 8B.2C12 monoclonal antibody reacts with mouse CD366 (TIM3), a Th1-specific cell surface protein. CD366 is a type I transmembrane protein and contains an immunoglobulin and a mucin-like domain in its extracellular portion and a tyrosine phosphorylation motif in its cytoplasmic portion. CD366 is expressed selectively by differentiated CD4+ Th1 and CD8+ Tc1, but is absent on Th2 and Tc2. Other hematopoietic cell types, including naïve T cells, B cells, macrophages and dendritic cells, do not express CD366, at least at the protein level. Expression of CD366 is upregulated at a late stage of T cell differentiation on Th1 cells after 3 rounds of in vitro polarization suggesting a role for this molecule in the transport or effector function of Th1 cells rather than a contribution to T cell differentiation. In an experimental autoimmune encephalomyelitis (EAE) model, CD366 was shown to be expressed on most CD4+ and CD8+ T cells in the central nervous system at the onset of clinical signs of disease, while less than 2% of CD4+ cells in the periphery expressed CD366 after immunization. In this model, in vivo administration of 8B.2C12 resulted in a hyperacute and atypical disease phenotype. It is postulated that the engagement of CD366 during T cell activation results in the expansion and activation of macrophages and increased severity of autoimmune disease. The Tim gene family may have an important role in the regulation of autoimmunity and allergies.

The 8B.2C12 antibody binds to the BALB/c allele of CD366 while reactivity to the C57Bl/6 allele is significantly weaker.
Description & Specifications

Specifications

CD366 (TIM3)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
8B.2C12
TIM-3, HAVCR2, T cell immunoglobulin domain, mucin-like domain
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL