missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD366 (TIM3) Mouse anti-Human, PerCP-eFluor 710, Clone: F38-2E2, eBioscience™

Mouse Monoclonal Antibody

$205.90 - $487.20

Specifications

Antigen CD366 (TIM3)
Clone F38-2E2
Host Species Murine
Gene Alias HAVCR2, T cell immunoglobulin and mucin domain-containing protein 3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245896 Life Technologies
LS46310941
25 Tests Each for $205.90
50245897 Life Technologies
LS46310942
100 Tests Each for $487.20
Specifications

Description

This F38-2E2 monoclonal antibody reacts with human CD366, also known as T cell immunoglobulin and mucin domain-containing protein 3 (TIM3) or HAVCR2. This cell surface receptor is expressed on activated CD4+ T cell subsets (e.g. Th1, Th17, and Treg), CD8+ T cells, monocytes, dendritic cells, and mast cells. Due to alternative splicing, CD366 exists as membrane-bound and soluble forms. Galectin-9 has been identified as the ligand for CD366. In humans, this receptor negatively regulates CD4+ T cells, influencing the secretion of some Th1- and Th17-related cytokines. CD366 has also been implicated in tolerance, autoimmune disease (e.g. multiple sclerosis), and HIV infection.
Description & Specifications

Specifications

CD366 (TIM3)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
F38-2E2
HAVCR2, T cell immunoglobulin and mucin domain-containing protein 3
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.06μg)/test