CD366 (TIM3), PE-Cyanine7, clone: F38-2E2, eBioscience™
Mouse Monoclonal Antibody
Manufacturer:  Life Technologies LS25310942
 View more versions of this product
Catalog No. 501128674
Description
Description
HAVCR2 is a 281 A.A Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. It consists of an IgV-like domain and a mucin-like domain in the extracellular region and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. HAVCR2 plays an important role in the regulation of macrophage activation and induction of autoimmune diseases. It down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of HAVCR2 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease.
00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. TIM3 (Hepatitis A virus cellular receptor 2, HAVCR2, T-cell immunoglobulin, mucin-dmain containing-3) is a 281 amino acid long, Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. TIM3 consists of an IgV-like domain, a mucin-like domain in the extracellular region, and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. TIM3 is involved in macrophage activation and induction of autoimmune diseases. Further, TIM3 down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of TIM3 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease, and also enhances the severity of experimental autoimmune encephalomyelitis in mice.
Specifications
Specifications
CD366 (TIM3)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q8TDQ0
HAVCR2,TIM-3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
F38-2E2
PE-Cyanine7
HAVCR2
Liquid
HAVCR2
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
84868
Documents
For Research Use Only.