CD366 (TIM3), PE, clone: F38-2E2, eBioscience™
Mouse Monoclonal Antibody
Manufacturer:  Life Technologies LS12310942
 View more versions of this product
Catalog No. 501129096
Description
Description
HAVCR2 is a 281 A.A Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. It consists of an IgV-like domain and a mucin-like domain in the extracellular region and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. HAVCR2 plays an important role in the regulation of macrophage activation and induction of autoimmune diseases. It down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of HAVCR2 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease.
Description: This F38-2E2 monoclonal antibody reacts with human CD366, also known as T cell immunoglobulin and mucin domain-containing protein 3 (TIM3) or HAVCR2. This cell surface receptor is expressed on activated CD4+ T cell subsets (e.g. Th1, Th17, and Treg), CD8+ T cells, monocytes, dendritic cells, and mast cells. Due to alternative splicing, CD366 exists as membrane-bound and soluble forms. Galectin-9 has been identified as the ligand for CD366. In humans, this receptor negatively regulates CD4+ T cells, influencing the secretion of some Th1- and Th17-related cytokines. CD366 has also been implicated in tolerance, autoimmune disease (e.g. multiple sclerosis), and HIV infection.Applications Reported: This F38-2E2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This F38-2E2 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications
Specifications
CD366 (TIM3)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q8TDQ0
HAVCR2,TIM-3
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
F38-2E2
PE
HAVCR2
Liquid
HAVCR2
IgG1, kappa, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
84868
Documents
For Research Use Only.