Please login to your online account to display your discounted pricing

CD363 Mouse anti-Human, eFluor 660, Clone: SW4GYPP, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $575.65

Specifications

Antigen CD363
Clone SW4GYPP
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124346

 
life technologies
LS50363941
25 tests Each for $220.40

501124347

 
life technologies
LS50363942
100 tests Each for $575.65
Description & Specifications

Specifications

Antigen CD363
Clone SW4GYPP
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The SW4GYPP monoclonal antibody recognizes human CD363, also known as sphingosine-1-phosphate receptor 1 (S1PR1, S1P1), or endothelial differentiation G-protein coupled receptor 1 (EDG1). CD363 is a part of a family of five G-protein-coupled receptors that bind sphingosine-1-phosphate (S1P), a lysophospholipid mediator. It has been shown to be primarily expressed by lymphocytes but is also seen on endothelial cells, B cells, NK cells, monocytes/macrophages, neural progenitors, neurons, astrocytes, and oligodendrocytes. CD363 surface expression is dependent on the extracellular concentration of S1P and its signaling is important for lymphocyte egress from secondary lymphoid tissues and the thymus. CD363 plays a key role in many cellular processes like angiogenesis, vascular maturation, endothelial barrier function and trafficking of immune cells. Therapeutic CD363 modulators have been successful in the treatment of relapsing-remitting multiple sclerosis through sequestration of immune cells in secondary lymphoid organs and, therefore, away from sites of inflammation.