Please login to your online account to display your discounted pricing

CD360 (IL-21 Receptor) Mouse anti-Human, PE, Clone: 2SX21R, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen CD360 (IL-21 Receptor)
Clone 2SX21R
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122311

 
life technologies
LS12360141
25 Tests Each for $172.55

501122312

 
life technologies
LS12360142
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen CD360 (IL-21 Receptor)
Clone 2SX21R
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

This 2SX21R monoclonal antibody reacts with human CD360 (Interleukin-21 Receptor, IL-21R). IL-21R forms a heterodimeric receptor complex with the common gamma chain to form a functional receptor for IL-21. IL-21 is a class I cytokine of the common gamma chain family of cytokines and signals through the IL-21R complex to activate the JAK/STAT (Janus Kinase/Signal Transducers and Activators of Transcription) signaling pathway. Activation of the JAK/STAT pathway by IL-21 promotes transcription of genes responsive to STAT1 and STAT3. IL-21 is mainly produced by CD4+ T cells and mediates the proliferation, differentiation, and effector functions of cells expressing IL-21R such as B cells, T cells, NK cells, dendritic cells and other populations of myeloid cells. IL-21 and its receptor are implicated in the pathogenesis of chronic viral infections, autoimmunity, allergy, organ transplantation, and cancer.