Please login to your online account to display your discounted pricing

Alfa Aesar™ CD36 Peptide P (93-110)

For cellular and molecular biology applications

Specifications

Product Type CD36 Peptide P (93-110)
Molecular Weight 2270.45
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Tyr-Arg-Val-Arg-Phe-Leu-Ala-Lys-Glu-Asn-Val-Thr-Gln-Asp-Ala-Glu-Asp-Asn-Cys
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66970EXD

 
alfa aesar™
J66970EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66970MCR

 
alfa aesar™
J66970MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type CD36 Peptide P (93-110)
Molecular Weight 2270.45
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Tyr-Arg-Val-Arg-Phe-Leu-Ala-Lys-Glu-Asn-Val-Thr-Gln-Asp-Ala-Glu-Asp-Asn-Cys
Regulatory Status RUO
Purification Method HPLC