Please login to your online account to display your discounted pricing

Alfa Aesar™ CD36 Peptide P (93-110)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66970MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66970MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type CD36 Peptide P (93-110)
Storage Requirements -30° to -10°C
Sequence Tyr-Arg-Val-Arg-Phe-Leu-Ala-Lys-Glu-Asn-Val-Thr-Gln-Asp-Ala-Glu-Asp-Asn-Cys
Molecular Weight 2270.45
Purification Method HPLC
Format Lyophilized powder
Regulatory Status RUO
Quantity 1mg