Please login to your online account to display your discounted pricing

Alfa Aesar™ CD36 Peptide P (139-155)

For cellular and molecular biology applications

Specifications

Product Type CD36 Peptide P (139-155)
Molecular Weight 1976.19
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Cys-Asn-Leu-Ala-Val-Ala-Ala-Ala-Ser-His-Ile-Tyr-Gln-Asn-Gln-Phe-Val-Gln
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66927EXD

 
alfa aesar™
J66927EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66927MCR

 
alfa aesar™
J66927MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type CD36 Peptide P (139-155)
Molecular Weight 1976.19
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Cys-Asn-Leu-Ala-Val-Ala-Ala-Ala-Ser-His-Ile-Tyr-Gln-Asn-Gln-Phe-Val-Gln
Regulatory Status RUO
Purification Method HPLC