missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD36 Mouse anti-Human, FITC, Clone: eBioNL07 (NL07), eBioscience™

Mouse Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen CD36
Clone eBioNL07 (NL07)
Host Species Murine
Gene Alias GP IIIb, Thrombospondin Receptor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509536 Life Technologies
LS11036942
100 Tests Each for $288.55
509535 Life Technologies
LS11036941
25 Tests Each for $143.55
Specifications

Description

The monoclonal antibody eBioNL07 recognizes human CD36, which is a member of the class B scavenger receptor family. CD36 was originally identified as a platelet-membrane glycoprotein also called glycoprotein IV and a receptor for thrombospondin-1 (TSP-1) and extracellular matrix proteins. Binding to TSP-1 is in the CLESH (CD36 LIMP-II Emp sequence homology) domain of CD36. CD36 expression is broad and includes microvascular (but not large vessel) endothelium, adipocytes, skeletal muscle, dendritic cells, epithelia of the retina, breast, and intestine, smooth muscle cells, and hematopoietic cells, including erythroid precursors, platelets, monocytes/macrophages, DCs and megakaryocytes. Expression on platelets is absent on Nak-a negative donors. Unlike other scavenger receptor, CD36 binds LDL that has been exposed to “minimally” oxidizing conditions. CD36 is also a fatty acid translocase (FAT) necessary for the transport of long-chain fatty acids (LCFAs) and therefore may play a role in atherosclerosis.
Description & Specifications

Specifications

CD36
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioNL07 (NL07)
GP IIIb, Thrombospondin Receptor
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test