missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD357 (GITR) Rat anti-Mouse, Clone: DTA-1, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$183.48 - $500.28

Specifications

Antigen CD357 (GITR)
Clone DTA-1
Host Species Rat
Gene Alias TNFRSF18, AITR, glucocorticoid-induced TNFR-related
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122962 Life Technologies
LS16587483
200μg Each for $500.28
5014587 Life Technologies
LS16587481
50μg Each for $183.48
Specifications

Description

The DTA-1 monoclonal antibody reacts with mouse GITR, Glucocorticoid-Induced TNFR family gene, also known as TNFRSF18. In naïve mice, GITR is expressed predominantly by CD4+CD25+ T regulatory cells (Treg) and by CD25+ CD4+ CD8- thymocytes. Stimulation of GITR with DTA-1 antibody abrogates Treg cell-mediated suppression. Although it does not deplete CD357 expressing cells in vivo, it has been shown to induce tumorogenesis. The removal of GITR-expressing Treg cells or the administration of DTA-1 resulted in organ specific autoimmune disease.
Description & Specifications

Specifications

CD357 (GITR)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, no sodium azide
DTA-1
TNFRSF18, AITR, glucocorticoid-induced TNFR-related
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL