missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD357 (AITR/GITR) Mouse anti-Human, PE-eFluor 610, Clone: eBioAITR, eBioscience™

Mouse Monoclonal Antibody

$187.44 - $469.92

Specifications

Antigen CD357 (AITR/GITR)
Clone eBioAITR
Host Species Murine
Gene Alias TNFRSF18, activation-inducible TNFR family member
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124841 Life Technologies
LS61587541
25 Tests Each for $187.44
501128618 Life Technologies
LS61587542
100 Tests Each for $469.92
Specifications

Description

The monoclonal antibody reacts with human AITR, Activation Inducible TNFR family member, an approximately 25kDa member of the TNFR superfamily. AITR mRNA is detected in lymph node, peripheral blood leukocytes and weakly in spleen. AITR mRNA expression was upregulated within 24 hour on PMA/ionomycin or PHA stimulated PBMC. At the protein level, AITR is expressed by a small population of activated PBMC. AITR associates with TRAF1, TRAF2 and TRAF3 and induces nuclear factor NF-kappaB activation via TRAF2. Recently TL6 (AITRL) has been reported as the ligand for AITR. Interaction of AITR with AITRL is important for cross-talk between T lymphocytes and endothelial cells.
Description & Specifications

Specifications

CD357 (AITR/GITR)
Murine
Flow Cytometry
PE-eFluor 610
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (1.0μg)/test
eBioAITR
TNFRSF18, activation-inducible TNFR family member
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer